Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dioxan-2-one 98.0+%, TCI America™
SDP

Catalog No. D464425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D464425G 25 g
D46445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D464425G Supplier TCI America Supplier No. D464425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3041-16-5
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
MDL Number MFCD00215855
InChI Key VPVXHAANQNHFSF-UHFFFAOYSA-N
Synonym p-Dioxanone
PubChem CID 18233
IUPAC Name 1,4-dioxan-2-one
SMILES C1COC(=O)CO1

Specifications

Melting Point 28°C
Color White
Quantity 25 g
Physical Form Crystal-Lump at 20°C
Boiling Point 215°C
Formula Weight 102.09
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1,4-Dioxan-2-one
RTECSNumber JH2290000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.