Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dioxane, 99.8%, Extra Dry, Stabilized, Acros Organics™

Click to view available options
Quantity:
100mL
Packaging:
AcroSeal™ Glass Bottle
123-91-1
C4H8O2
88.106
MFCD00006571
RYHBNJHYFVUHQT-UHFFFAOYSA-N
dioxane,p-dioxane,1,4-diethylene dioxide,diethylene ether,dioxan,1,4-dioxacyclohexane,diethylene dioxide,dioxanne,di ethylene oxide,tetrahydro-p-dioxin
31275
CHEBI:47032
1,4-dioxane
C1COCCO1
123-91-1
C4H8O2
88.106
MFCD00006571
RYHBNJHYFVUHQT-UHFFFAOYSA-N
dioxane,p-dioxane,1,4-diethylene dioxide,diethylene ether,dioxan,1,4-dioxacyclohexane,diethylene dioxide,dioxanne,di ethylene oxide,tetrahydro-p-dioxin
31275
CHEBI:47032
1,4-dioxane
C1COCCO1
Appearance Colorless
Assay Percent Range 99.8%
UN Number 1165
Merck Index 153,328
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone, benzene, toluene,, ether and other common organic solvents
Formula Weight 88.11
Percent Purity 99.8%
Grade Extra Dry
Density 1.0340g/mL
Density at 25 degC 1.034g/cm3
Ignition Point 355°C (671°F)
Odor Petroleum distillates
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4205 to 1.4220
Solubility in Water Yes
Solubility Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone, benzene, toluene,, ether and other common organic solvents
Specific Gravity 1.034
Vapor Pressure 41mbar at 20°C
Flash Point 12°C
Viscosity 1.32 mPa.s (20°C)
Physical State Liquid
Boiling Point 101.0°C
Color Colorless
Melting Point 12.0°C
pH 6 to 8
Quantity 100mL
Avg. Mol. Wt. or Mol. Wt. Range >95%
Chemical Name or Material 1,4-Dioxane
Assay Extra Dry, Stabilized
Show More Show Less

GHS S Statement

  • Causes serious eye irritation
  • H351: Suspected of causing cancer
  • H335: May cause respiratory irritation
  • H225: Highly flammable liquid and vapor
  • EUH019: May form explosive peroxides

GHS P Statement

  • Keep away from heat/sparks/open flames/hot surfaces.
  • Keep container tightly closed
  • Avoid breathing dust/fume/gas/mist/vapors/spray
  • Wear protective gloves/protective clothing/eye protection/face protection
  • IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

Danger

EINECSNumber : 204-661-8