Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dioxane in Dimethyl Sulfoxide, SPEX CertiPrep™
SDP

Catalog No. USPS1715DMS Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USPS1715DMS 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USPS1715DMS Supplier SPEX CertiPrep Supplier No. USPS1715DMSO

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,4-Dioxane123-91-11900μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Packaging 2mL Amber Prescored Glass Ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Explosion Limit 1.8% (volume); 63% (volume)
Ignition Point 270°C
Vapor Pressure 2.5 hPA (20°C)
Analyte or Component Names 1,4-Dioxane
Density 1.1g/cm3
Physical Form Fluid
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Element 1,4-Dioxane
Solvent or Matrix Dimethyl Sulfoxide
Quantity 1 mL
Grade USP
Concentration or Composition (by Analyte or Components) 1900μg/mL
Chemical Name or Material 1,4-Dioxane in Dimethyl Sulfoxide, USP 467 Standard
Show More Show Less
DOTInformation Nonhazardous
Recommended Storage Freezer

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.