Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dioxane, HPLC Grade, 99% min

Catalog No. AA22913K7
Click to view available options
Quantity:
1 L
4 L
4 x 1 L

CAS: 123-91-1 | C4H8O2 | 88.106 g/mol

1,4-Dioxane HPLC Grade is used as a mobile phase in high performance liquid chromatography and liquid chromatography coupled with mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00006571
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Melting Point 11.8°C
Color Colorless
Appearance Colorless liquid
Density 1.034 g/cm3 at 20°C
Boiling Point 100°C to 102°C
Quantity 4 L
Flash Point 12°C (54°F)
Ignition Point 375°C (707°F)
Odor Ether like
Physical Form Liquid
Refractive Index 1.422
Assay Percent Range 99% min.
Vapor Pressure 41 hPa (31mm Hg) at 20°C
UN Number 1165
Beilstein 102551
Solubility Information Miscible with water,most organic solvents,aromatic hydrocarbons and oils.
Formula Weight 88.11
Grade HPLC
Concentration or Composition (by Analyte or Components) 1, 4-Dioxane
Chemical Name or Material 1, 4-Dioxane, Filtered through 0.2μm filters.
Show More Show Less
Hazard Category H225-H319-H335-H336-H350-H373
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P370+P378q-P501c
DOTInformation DOT Class: 3, Packaging Group: II
EINECSNumber 204-661-8
RTECSNumber JG8225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.