Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-dioxaspiro[4.5]decan-8-one, Thermo Scientific™

Catalog No. SB00697EB
Click to view available options
Quantity:
25 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 4746-97-8
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.181
InChI Key VKRKCBWIVLSRBJ-UHFFFAOYSA-N
Synonym 1,4-dioxaspiro 4.5 decan-8-one, 1,4-cyclohexanedione monoethylene acetal, 1,4-cyclohexanedione monoethylene ketal, 1,4-cyclohexanedione monoethyleneacetal, 1,4-dioxa-spiro 4.5 decan-8-one, 1,4-cyclohexanedione monoethyleneketal, cyclohexane-1,4-dione monoethylene ketal, 1,4-dioxaspiro 4.5 decane-8-one
PubChem CID 567415
IUPAC Name 1,4-dioxaspiro[4.5]decan-8-one
SMILES C1CC2(CCC1=O)OCCO2

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 156.18
Chemical Name or Material 1, 4-Dioxaspiro[4.5]decan-8-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.