Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Diphenyl-1-butanone, 98%, Thermo Scientific™

Catalog No. AC232820010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC232820010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC232820010 Supplier Thermo Scientific Chemicals Supplier No. 232820010

Chemical Identifiers

CAS 5407-91-0
Molecular Formula C16H16O
Molecular Weight (g/mol) 224.3
MDL Number MFCD00022005
InChI Key GBUMEGLMTNAXOM-UHFFFAOYSA-N
Synonym 4-phenylbutyrophenone, 1,4-diphenyl-1-butanone, 1-butanone, 1,4-diphenyl, 4-phenylbutyrophenon, pubchem8921, 1-butanone,4-diphenyl, 1,4-diphenyl-butan-1-one, 1-butanone,1,4-diphenyl, 3-phenylpropyl phenyl ketone, acmc-1am65
PubChem CID 79413
IUPAC Name 1,4-diphenylbutan-1-one
SMILES C1=CC=C(C=C1)CCCC(=O)C2=CC=CC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 55°C to 59°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CO(CH2)3C6H5
Quantity 1 g
Formula Weight 224.3
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1, 4-Diphenyl-1-butanone, 98%
Show More Show Less
EINECSNumber 226-471-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.