Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Diphenylbutadiyne, 99%

Catalog No. AAA1347114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1347114 25 g
AAA1347122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1347114 Supplier Thermo Scientific Chemicals Supplier No. A1347114
Add to Cart

CAS: 886-66-8 | C16H10 | 202.26 g/mol

1,4-Diphenylbutadiyne was used in the preparation of 7,8-dehydropurpurin dimers via two-fold Pd-catalyzed [3+2] annulation of meso-bromoporphyrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Diphenylbutadiyne was used in the preparation of 7,8-dehydropurpurin dimers via two-fold Pd-catalyzed [3+2] annulation of meso-bromoporphyrin.

Solubility
Sparingly soluble in water(5.5E-4 g/L) (25°C). Soluble in toluene almost transparency.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886-66-8
Molecular Formula C16H10
Molecular Weight (g/mol) 202.26
MDL Number MFCD00004787
InChI Key HMQFJYLWNWIYKQ-UHFFFAOYSA-N
Synonym 1,4-diphenylbutadiyne, diphenylbutadiyne, diphenyldiacetylene, butadiyne, diphenyl, diphenyl-1,3-butadiyne, benzene, 1,1'-1,3-butadiyne-1,4-diyl bis, 1,4-diphenyl-1,3-butadiyne, 1,4-diphenylbuta-1,3-diyne, 1,1'-buta-1,3-diyne-1,4-diyldibenzene, 4-phenyl-1,3-butadiynyl benzene
PubChem CID 70174
ChEBI CHEBI:51588
IUPAC Name 4-phenylbuta-1,3-diynylbenzene
SMILES C1=CC=C(C=C1)C#CC#CC1=CC=CC=C1

Specifications

Melting Point 86°C to 87°C
Quantity 25 g
Beilstein 1910105
Solubility Information Sparingly soluble in water(5.5E-4g/L) (25°C). Soluble in toluene almost transparency.
Formula Weight 202.26
Percent Purity 99%
Chemical Name or Material 1,4-Diphenylbutadiyne
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-953-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.