Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1,4-Dithioerythritol, 98+%
Click to view available options
Quantity:
5 g
25 g
Description
Reagent for maintaining -SH groups in the reduced state, quantitatively reduces disulfides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reagent for maintaining -SH groups in the reduced state, quantitatively reduces disulfides.
Solubility
Soluble in water (15.4 mg/ml), and methanol.
Notes
Air Sensitive, store away form air. Keep the container tightly sealed. Store away from reducing agent. Store under dry inert gas.
Reagent for maintaining -SH groups in the reduced state, quantitatively reduces disulfides.
Solubility
Soluble in water (15.4 mg/ml), and methanol.
Notes
Air Sensitive, store away form air. Keep the container tightly sealed. Store away from reducing agent. Store under dry inert gas.

Specifications
Specifications
Melting Point | 82°C to 84°C |
Odor | Stench |
Quantity | 5 g |
UN Number | UN3335 |
Beilstein | 1719756 |
Sensitivity | Air sensitive |
Solubility Information | Soluble in water (15.4mg/ml),and methanol. |
IUPAC Name | (2R,3S)-1,4-bis(sulfanyl)butane-2,3-diol |
Formula Weight | 154.25 |
Percent Purity | 99% |
Show More |
Safety and Handling
Hazard Category | H302-H315-H319-H335-H500 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
DOTInformation | Hazard Class: 9; Packaging Group: III |
EINECSNumber | 229-998-8 |
RTECSNumber | KF2410000 |
TSCA | Yes |
Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction