Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Fluoro-2-nitrophenyl)azepane, 97%, Thermo Scientific™

Catalog No. AAH6321006
Click to view available options
Quantity:
250 mg
1 g
5 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 897598-47-9
Synonym 1-(4-Fluoro-2-nitrophenyl)hexamethyleneimine

Specifications

Quantity 5 g
UN Number UN3259
Formula Weight 238.25
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(4-Fluoro-2-nitrophenyl)azepane
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.