Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-(4-Fluorophenyl)-2-propylamine, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is employed as intermediate for drug discovery.
Solubility
Soluble in ethanol, dimethyl sulfoxide and dimethyl formamide.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
It is employed as intermediate for drug discovery.
Solubility
Soluble in ethanol, dimethyl sulfoxide and dimethyl formamide.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Specifications
Specifications
Quantity | 1 g |
Solubility Information | Soluble in ethanol,dimethyl sulfoxide and dimethyl formamide. |
Formula Weight | 153.2 |
Percent Purity | 97% |
Chemical Name or Material | 1-(4-Fluorophenyl)-2-propylamine |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
RTECSNumber | SH7040000 |
TSCA | No |
Recommended Storage | Ambient temperatures |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction