Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Hydrazinobenzyl)-1H-1,2,4-triazole Dihydrochloride 97.0+%, TCI America™
SDP

Catalog No. H1467200MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg

Specifications

CAS 212248-62-9
Molecular Formula C9H11N5
Quantity 200 mg
MDL Number MFCD06795497
Synonym 4-[(1,2,4-Triazol-1-yl)methyl]phenylhydrazine Dihydrochloride
InChI Key FUJKUTJPEGEMHG-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1CN2C=NC=N2)NN
IUPAC Name [4-(1,2,4-triazol-1-ylmethyl)phenyl]hydrazine
Molecular Weight (g/mol) 189.222
PubChem CID 7472049
Formula Weight 262.14
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Hydrazinobenzyl)-1H-1,2,4-triazole Dihydrochloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.