Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Methoxybenzyl)piperazine, 97%

Catalog No. AAH5509903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5509903 1 g
AAH5509906 5 g
AAH5509914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5509903 Supplier Thermo Scientific Chemicals Supplier No. H5509903

missing translation for 'validMainframeMsg'

CAS: 21867-69-6 | C12H20N2O | 208.30 g/mol

1-(4-Methoxybenzyl)piperazine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(4-Methoxybenzyl)piperazine is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21867-69-6
Molecular Formula C12H20N2O
Molecular Weight (g/mol) 208.30
MDL Number MFCD01075236
InChI Key MGLUVVBFISROAH-UHFFFAOYSA-P
PubChem CID 231905
IUPAC Name 1-[(4-methoxyphenyl)methyl]piperazine
SMILES COC1=CC=C(C[NH+]2CC[NH2+]CC2)C=C1

Specifications

Melting Point 32°C to 36°C
Color White to Yellow
Flash Point >110°C (230°F)
Quantity 1 g
Synonym 1-4-methoxybenzyl piperazine, 1-4-methoxyphenyl methyl piperazine, piperazine,1-4-methoxyphenyl methyl, 1-4-methoxy-benzyl-piperazine, piperazine, 1-4-methoxyphenyl methyl, 4-methoxy-1-piperazinylmethyl benzene, 1-4-methoxybenzyl-piperazine, 1-4-methoxybenzyl piperazine #, piperazine, 1-4-methoxybenzyl, 1-4-methoxy-phenylmethyl piperazine
Formula Weight 206.29
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(4-Methoxybenzyl)piperazine
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.