Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Methoxyphenyl)piperazine, 96%, Thermo Scientific™

Catalog No. AC255830250
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 38212-30-5
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.26
MDL Number MFCD00040736
InChI Key MRDGZSKYFPGAKP-UHFFFAOYSA-N
Synonym 1-4-methoxyphenyl piperazine, n-4-methoxyphenyl piperazine, n-p-methoxyphenyl piperazine, piperazine, 1-4-methoxyphenyl, unii-p385m92gyg, 4-methoxyphenylpiperazine, 1-4-anisyl piperazine, chembl59517, 1-4-methoxyphenyl-piperazine, 1-4-methoxy-phenyl-piperazine
PubChem CID 269722
IUPAC Name 1-(4-methoxyphenyl)piperazine
SMILES COC1=CC=C(C=C1)N1CCNCC1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Amber to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 192.25
Percent Purity 96%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 1-(4-Methoxyphenyl)piperazine, 96%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 253-829-7
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.