Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Nitrophenyl)-2-piperidone 98.0+%, TCI America™
SDP

Catalog No. N09751G
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 38560-30-4
Molecular Formula C11H12N2O3
Molecular Weight (g/mol) 220.228
MDL Number MFCD04037154
InChI Key VPCQXIGQMFWXII-UHFFFAOYSA-N
PubChem CID 22324463
IUPAC Name 1-(4-nitrophenyl)piperidin-2-one
SMILES C1CCN(C(=O)C1)C2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 99°C
Color Yellow
Quantity 1 g
Formula Weight 220.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Nitrophenyl)-2-piperidone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.