Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanidine nitrate, 98%, Thermo Scientific™

Catalog No. AC215310025
Click to view available options
Quantity:
2.5 kg

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-93-4
Molecular Formula CH6N4O3
Molecular Weight (g/mol) 122.084
MDL Number MFCD00013028
InChI Key CNUNWZZSUJPAHX-UHFFFAOYSA-N
Synonym guanidine nitrate, guanidinium nitrate, guanidine; nitric acid, guanidine, mononitrate, guanidine, nitrate, guanidine mononitrate, guanidine nitrate 1:1, guanidinenitrate, pubchem21372, acmc-209kpa
PubChem CID 10481
IUPAC Name guanidine;nitric acid
SMILES C(=N)(N)N.[N+](=O)(O)[O-]

Specifications

Melting Point 212°C to 215°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula H2NC(=NH)NH2·HNO3
Quantity 2.5 kg
Beilstein 03, 86
Merck Index 15, 4597
Formula Weight 122.08
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material Guanidine nitrate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-060-1
RTECSNumber MF4350000
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.