Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Phenylenediacetonitrile, 97%

Catalog No. aal1346406
Click to view available options
Quantity:
5 g
25 g

CAS: 622-75-3 | C10H8N2 | 156.188 g/mol

1,4-Phenylenediacetonitrile, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Phenylenediacetonitrile, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-75-3
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.188
MDL Number MFCD00001923
InChI Key FUQCKESKNZBNOG-UHFFFAOYSA-N
Synonym 1,4-phenylenediacetonitrile, 1,4-benzenediacetonitrile, p-xylylene dicyanide, p-phenylenediacetonitrile, p-benzenediacetonitrile, 2,2'-1,4-phenylene diacetonitrile, p-bis cyanomethyl benzene, 1,4-bis cyanomethyl benzene, p-cyanomethyl benzyl cyanide, 2-4-cyanomethyl phenyl acetonitrile
PubChem CID 69327
IUPAC Name 2-[4-(cyanomethyl)phenyl]acetonitrile
SMILES C1=CC(=CC=C1CC#N)CC#N

Specifications

Melting Point 94°C to 98°C
Quantity 5 g
UN Number UN3439
Beilstein 1866157
Solubility Information Insoluble in water.
Formula Weight 156.19
Percent Purity 97%
Chemical Name or Material 1,4-Phenylenediacetonitrile
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement GHS P Statement
P280h-P309-P310
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 210-751-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.