Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Pyridyl)piperazine, 97%

Catalog No. aal1939606
Click to view available options
Quantity:
5 g
25 g

CAS: 1008-91-9 | C9H13N3 | 163.224 g/mol

1-(4-Pyridyl)piperazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is also used to produce 1-pyridin-4-yl-4-(4-vinyl-benzyl)-piperazine with 1-(chloromethyl)-4-vinylbenzene by heating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(4-Pyridyl)piperazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is also used to produce 1-pyridin-4-yl-4-(4-vinyl-benzyl)-piperazine with 1-(chloromethyl)-4-vinylbenzene by heating.

Solubility
Soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Store under inert gas. It is sensitive to air. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1008-91-9
Molecular Formula C9H13N3
Molecular Weight (g/mol) 163.224
MDL Number MFCD00040745
InChI Key OQZBAQXTXNIPRA-UHFFFAOYSA-N
Synonym 1-4-pyridyl piperazine, 1-pyridin-4-yl piperazine, 1-pyridin-4-yl-piperazine, 4-piperazinopyridine, 4-pyridylpiperazine, 1-4-pyridyl-piperazine, 4-1-piperazino pyridine, 1-4-pyridinyl piperazine, piperazine, 1-4-pyridinyl
PubChem CID 70517
IUPAC Name 1-pyridin-4-ylpiperazine
SMILES C1CN(CCN1)C2=CC=NC=C2

Specifications

Melting Point 136°C to 141°C
Boiling Point 195°C to 200°C (0.3 mmHg)
Quantity 5 g
UN Number UN3259
Beilstein 609705
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 163.22
Percent Purity 97%
Chemical Name or Material 1-(4-Pyridyl)piperazine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 213-764-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.