Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Diaminonaphthalene 98.0+%, TCI America™
SDP

Catalog No. D0101100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0101100G 100 g
D010125G 25 g
D0101500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0101100G Supplier TCI America Supplier No. D0101100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2243-62-1
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.204
MDL Number MFCD00004029
InChI Key KQSABULTKYLFEV-UHFFFAOYSA-N
Synonym 1,5-diaminonaphthalene, 1,5-naphthalenediamine, 1,5-naphthylenediamine, 1,5-napthalenediamine, unii-13pd3j52lk, ccris 422, 1,5-diamino naphthalene, 1,5-diaminonaphtalene, dsstox_cid_916, acmc-1crp7
PubChem CID 16720
ChEBI CHEBI:53003
IUPAC Name naphthalene-1,5-diamine
SMILES C1=CC2=C(C=CC=C2N)C(=C1)N

Specifications

Melting Point 187°C
Boiling Point 210°C
Quantity 100 g
UN Number 3077
Formula Weight 158.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Diaminonaphthalene
EINECSNumber (4)-0323
RTECSNumber QJ3400000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.