Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1,5-Diaminopentane, 98%
Click to view available options
Quantity:
5 g
25 g
Description
1,5-Diaminopentane is mainly used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,5-Diaminopentane is mainly used in organic synthesis.
Solubility
Soluble in water.
Notes
Air sensitive & hygroscopic. Incompatible with acid chlorides, acids, acid anhydrides, strong oxidizing agents, carbon dioxide.
1,5-Diaminopentane is mainly used in organic synthesis.
Solubility
Soluble in water.
Notes
Air sensitive & hygroscopic. Incompatible with acid chlorides, acids, acid anhydrides, strong oxidizing agents, carbon dioxide.
Specifications
Specifications
| Melting Point | 9°C |
| Density | 0.873 |
| Boiling Point | 178°C to 180°C |
| Assay Percent Range | 0.98 |
| Refractive Index | 1.4585 |
| Linear Formula | H2N(CH2)5NH2 |
| Quantity | 5 g |
| UN Number | UN2735 |
| Sensitivity | Air sensitive; Hygroscopic |
| Merck Index | 14,1609 |
| Show More |
Safety and Handling
| Hazard Category | H227-H302+H312-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318-H227 |
| Precautionary Statement | P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: POLYAMINES, LIQUID, CORROSIVE, N.O.S. |
| EINECSNumber | 207-329-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction