Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Dibromopentane, 97%

Catalog No. p-102544
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle

CAS: 111-24-0 | C5H10Br2 | 229.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-24-0
Molecular Formula C5H10Br2
Molecular Weight (g/mol) 229.94
MDL Number MFCD00000268
InChI Key IBODDUNKEPPBKW-UHFFFAOYSA-N
Synonym pentamethylene bromide, pentane, 1,5-dibromo, pentamethylene dibromide, unii-b8q228qys1, pentane-1,5-dibromide, 1,5-dibromo-pentane, 1,5-pentane dibromide, pentane,1,5-dibromo, br ch2 5br, ccris 8918
PubChem CID 8100
IUPAC Name 1,5-dibromopentane
SMILES C(CCBr)CCBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -34°C
Density 1.6800g/mL
Boiling Point 224°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.5112 to 1.5132
Linear Formula Br(CH2)5Br
Quantity 250 mL
Beilstein 01, 131
Specific Gravity 1.68
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene,chloroform,slightly soluble in carbon tetrachloride
Formula Weight 229.94
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1, 5-Dibromopentane, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-849-7
RTECSNumber SA0320000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.