Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Dichloro-1,1,3,3,5,5-hexamethyltrisiloxane 98.0+%, TCI America™
SDP

Catalog No. D282525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D282525G 25 g
D28255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D282525G Supplier TCI America Supplier No. D282525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3582-71-6
Molecular Formula C6H18Cl2O2Si3
Molecular Weight (g/mol) 277.363
MDL Number MFCD00045147
InChI Key GJIYNWRLGOMDEX-UHFFFAOYSA-N
Synonym 1,5-dichlorohexamethyltrisiloxane, 1,5-dichloro-1,1,3,3,5,5-hexamethyltrisiloxane, trisiloxane, 1,5-dichloro-1,1,3,3,5,5-hexamethyl, poly dimethylsiloxane , chlorine terminated, 2,6-dichloro-2,4,4,6-tetramethyl-3,5-dioxa-2,4,6-trisilaheptane, acmc-20ap1k, 1 5-dichlorohexamethyltrisiloxane, 1,5-dichloro-hexamethyltrisiloxane, 1,5-dichlorohexamethyl trisiloxane, trisiloxane,1,5-dichloro,hexamethyl
PubChem CID 77131
IUPAC Name bis[[chloro(dimethyl)silyl]oxy]-dimethylsilane
SMILES C[Si](C)(O[Si](C)(C)Cl)O[Si](C)(C)Cl

Specifications

Color Colorless
Boiling Point 184°C
Quantity 25 g
UN Number 2987
Formula Weight 277.36
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1,5-Dichloro-1,1,3,3,5,5-hexamethyltrisiloxane
EINECSNumber (7)-0472
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.