Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Dichloroanthraquinone 95.0+%, TCI America™
SDP

Catalog No. D032825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D032825G 25 g
D0328500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D032825G Supplier TCI America Supplier No. D032825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 82-46-2
Molecular Formula C14H6Cl2O2
Molecular Weight (g/mol) 277.1
MDL Number MFCD00001190
InChI Key MQIUMARJCOGCIM-UHFFFAOYSA-N
PubChem CID 6711
IUPAC Name 1,5-dichloroanthracene-9,10-dione
SMILES C1=CC2=C(C(=C1)Cl)C(=O)C3=C(C2=O)C(=CC=C3)Cl

Specifications

Melting Point 250°C
Quantity 25 g
Synonym 1,5-dichloroanthraquinone, 9,10-anthracenedione, 1,5-dichloro, 1,5-dichloro-9,10-anthraquinone, 1,5-dichloranthrachinon, unii-s335v6mf9e, anthraquinone, 1,5-dichloro, 1,5-dichloranthrachinon czech, 1, 5-dichloroanthraquinone, 1,5-dichloroanthra-9,10-quinone, 1,5 dichloro aq
Formula Weight 277.10
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Dichloroanthraquinone
EINECSNumber (4)-1246
RTECSNumber CB6495000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.