Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Dichloropentane, 98%

Catalog No. AAA1243336
Click to view available options
Quantity:
100 g
500 g

CAS: 628-76-2 | C5H10Cl2 | 141.035 g/mol

1,5-Dichloropentane is used in the preparation of symmetrical sulfide, tetrydrothiopyran (thiane) by reaction with sodium sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Dichloropentane is used in the preparation of symmetrical sulfide, tetrydrothiopyran (thiane) by reaction with sodium sulfide.

Solubility
Miscible with ethanol, ether and chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-76-2
Molecular Formula C5H10Cl2
Molecular Weight (g/mol) 141.035
MDL Number MFCD00001017
InChI Key LBKDGROORAKTLC-UHFFFAOYSA-N
Synonym pentamethylene chloride, pentane, 1,5-dichloro, pentamethylene dichloride, amylene chloride, unii-6n4a5v83we, 1,5-dichlorobutane, pentane,5-dichloro, acmc-1ay6s, 1,5-bis chloranyl pentane, wln: g5g
PubChem CID 12353
IUPAC Name 1,5-dichloropentane
SMILES C(CCCl)CCCl

Specifications

Melting Point -72°C
Density 1.103
Boiling Point 182°C
Flash Point 26°C (78°F)
Refractive Index 1.457
Linear Formula Cl(CH2)5Cl
Quantity 500 g
UN Number UN1152
Beilstein 1732340
Solubility Information Miscible with ethanol,ether and chloroform. Immiscible with water.
Formula Weight 141.04
Percent Purity 98%
Chemical Name or Material 1,5-Dichloropentane
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DICHLOROPENTANES
EINECSNumber 211-053-6
RTECSNumber SA0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.