Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Dimethyl-3-(trifluoromethyl)pyrazole 98.0+%, TCI America™
SDP

Catalog No. D50711G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50711G 1 g
D5071200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50711G Supplier TCI America Supplier No. D50711G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 79080-31-2
Molecular Formula C6H7F3N2
Molecular Weight (g/mol) 164.131
MDL Number MFCD04972844
InChI Key LGTRXZPAYGKNLP-UHFFFAOYSA-N
PubChem CID 12747525
IUPAC Name 1,5-dimethyl-3-(trifluoromethyl)pyrazole
SMILES CC1=CC(=NN1C)C(F)(F)F

Specifications

Color Yellow
Boiling Point 71°C
Quantity 1 g
UN Number 1993
Formula Weight 164.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,5-Dimethyl-3-(trifluoromethyl)pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.