Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Hexadiene Diepoxide 96.0+%, TCI America™
SDP

Catalog No. H095325G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g
25 g

Catalog No. H095325G

Supplier: TCI America H095325G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1888-89-7
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD01723447
InChI Key HTJFSXYVAKSPNF-UHFFFAOYSA-N
Synonym 1,5-hexadiene diepoxide, 1,2,5,6-diepoxyhexane, 1,2:5,6-diepoxyhexane, 2-2-oxiran-2-yl ethyl oxirane, ccris 3751, hexane, 1,2:5,6-diepoxy, 1,2-di oxiran-2-yl ethane, 1,6-diepoxyhexane, 1,2-bisoxiranylethane, 1,5,6-diepoxyhexane
PubChem CID 15903
IUPAC Name 2-[2-(oxiran-2-yl)ethyl]oxirane
SMILES C1C(O1)CCC2CO2

Specifications

Color Yellow
Boiling Point 188°C
Quantity 25 g
Formula Weight 114.14
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,5-Hexadiene Diepoxide
RTECSNumber MO1575000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.