Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One 1.5 mL Polypropylene Microcentrifuge Tubes

Catalog No. 07000243 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Assorted
Dark Brown
Natural
Closure Type:
Attached Cap
Cap
Flat Cap
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Type
07-000-243 Assorted Flat Cap
07-000-244 Natural Cap
07-000-298 Dark Brown Attached Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07-000-243 Supplier Greiner Bio-One Supplier No. 616281

For single use only. Greiner Bio-One Cell 1.5mL Polypropylene Microcentrifuge Tubes are free of heavy metal.

  • Material: Polypropylene
  • For single use only
  • Available in different colors
  • Free of detectable DNase/RNase, human DNA and pyrogens
  • Suitable for Eppendorf centrifuges and systems
TRUSTED_SUSTAINABILITY

Specifications

Color Assorted
No. per Pack 500
Volume (Metric) 1.5mL
Sterility Non-sterile
Graduated No
Closure Type Flat Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.