Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Pentanediol, 97%

Catalog No. p-7021597
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 111-29-5 | C5H12O2 | 104.15 g/mol

1,5-Pentanediol is used as a plasticizer in cellulose products and adhesives. It is used as a brake fluid additive. It reacts with 3,4-dihydro-2H-pyran to get 5-tetrahydropyran-2-yloxy-pentan-1-ol. It is also used to prepare polyesters for emulsifying agents and resin intermediates. Further, it is used in ink, toner and colorant products. In addition to this, it is used in brake fluid compositions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Pentanediol is used as a plasticizer in cellulose products and adhesives. It is used as a brake fluid additive. It reacts with 3,4-dihydro-2H-pyran to get 5-tetrahydropyran-2-yloxy-pentan-1-ol. It is also used to prepare polyesters for emulsifying agents and resin intermediates. Further, it is used in ink, toner and colorant products. In addition to this, it is used in brake fluid compositions.

Solubility
Miscible with water, methanol, ethanol and acetone.

Notes
Hygroscopic. Incompatible with acid chlorides, acid anhydrides, oxidizing agents, chloroformates and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-29-5
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00002978
InChI Key ALQSHHUCVQOPAS-UHFFFAOYSA-N
Synonym 1,5-pentanediol, 1,5-dihydroxypentane, pentylene glycol, pentamethylene glycol, 1,5-pentylene glycol, 1,5 pentanediol, 1,5-pentamethylene glycol, alpha,omega-pentanediol, unii-07uxz0scst, 1,5-pentandiol
PubChem CID 8105
IUPAC Name pentane-1,5-diol
SMILES OCCCCCO

Specifications

Melting Point -16°C
Density 0.992
Boiling Point 241°C to 242°C
Flash Point 129°C (264°F)
Refractive Index 1.45
Linear Formula HO(CH2)5OH
Quantity 100 g
Beilstein 1560130
Sensitivity Hygroscopic
Merck Index 14,7117
Solubility Information Miscible with water,methanol,ethanol and acetone.
Formula Weight 104.15
Percent Purity 97%
Chemical Name or Material 1,5-Pentanediol
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 203-854-4
RTECSNumber SA0480000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.