Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Pentanediol, 98%

Catalog No. AC129950010
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Metal drum

CAS: 111-29-5 | C5H12O2 | 104.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-29-5
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00002978
InChI Key ALQSHHUCVQOPAS-UHFFFAOYSA-N
Synonym 1,5-pentanediol, 1,5-dihydroxypentane, pentylene glycol, pentamethylene glycol, 1,5-pentylene glycol, 1,5 pentanediol, 1,5-pentamethylene glycol, alpha,omega-pentanediol, unii-07uxz0scst, 1,5-pentandiol
PubChem CID 8105
IUPAC Name pentane-1,5-diol
SMILES OCCCCCO

Specifications

Melting Point -18.0°C
Color Colorless
Density 0.9940g/mL
Boiling Point 242.0°C
Flash Point 135°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4490 to 1.4510
Linear Formula HO(CH2)5OH
Quantity 1 kg
Beilstein 01,481
Specific Gravity 0.994
Merck Index 15,7229
Solubility Information Solubility in water: miscible. Other solubilities: limited solubility in benzene, trichloroethylene,, petroleum ether, heptane, miscible with alcohol, acetone and ethyl acetate, 11 g/110g ether (25°C)
Viscosity 77 mPa.s (30°C)
Formula Weight 104.15
Percent Purity 98%
Physical Form Oily Liquid
Chemical Name or Material 1, 5-Pentanediol
Show More Show Less
EINECSNumber 203-854-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.