Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-[5-(Trifluoromethyl)-2-pyridyl]-1,4-diazepane,95+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 1 g |
| Formula Weight | 245.25 |
| Percent Purity | 97% |
| Chemical Name or Material | 1-[5-(Trifluoromethyl)-2-pyridyl]-1, 4-diazepane |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction