Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Bis(diphenylphosphino)hexane, 97%

Catalog No. p-7037006
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 19845-69-3 | C30H32P2 | 454.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19845-69-3
Molecular Formula C30H32P2
Molecular Weight (g/mol) 454.53
MDL Number MFCD00003053
InChI Key GPORFKPYXATYNX-UHFFFAOYSA-N
Synonym 1,6-bis diphenylphosphino hexane, 6-diphenylphosphanylhexyl diphenyl phosphane, hexamethylenebis diphenylphosphine, phosphine, 1,6-hexanediylbis diphenyl, phosphine,1,1'-1,6-hexanediyl bis 1,1-diphenyl, 6-diphenylphosphanyl hexyl diphenylphosphane, acmc-1ca9e, ksc491k2l, 1,6-bis diphenylphosino hexane
PubChem CID 2754312
IUPAC Name 6-diphenylphosphanylhexyl(diphenyl)phosphane
SMILES C(CCCP(C1=CC=CC=C1)C1=CC=CC=C1)CCP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 124°C to 126°C
Linear Formula (C6H5)2P(CH2)6P(C6H5)2
Quantity 1 g
Formula Weight 454.53
Percent Purity 97%
Chemical Name or Material 1,6-Bis(diphenylphosphino)hexane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.