Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Diaminohexane-N,N,N',N'-tetraacetic Acid 98.0+%, TCI America™
SDP

Catalog No. D201925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D201925G 25 g
D20195G 5 g
D201910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D201925G Supplier TCI America Supplier No. D201925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1633-00-7
Molecular Formula C14H24N2O8
Molecular Weight (g/mol) 348.35
MDL Number MFCD00004292
InChI Key YGDVXSDNEFDTGV-UHFFFAOYSA-N
Synonym 1,6-diaminohexane-n,n,n',n'-tetraacetic acid, hexamethylenediaminetetraacetic acid, hexamethylenediamine-n,n,n',n'-tetraacetic acid, 2-6-bis carboxymethyl amino hexyl carboxymethyl amino acetic acid, hdta, glycine,n,n'-1,6-hexanediylbis n-carboxymethyl, 6-bis carboxymethyl amino hexyl carboxymethyl amino acetic acid, glycine, n,n'-1,6-hexanediylbis n-carboxymethyl, 1,6-hexanediaminetetraacetic acid, 2,2',2,2'-hexane-1,6-diyldinitrilo tetraacetic acid
PubChem CID 74209
IUPAC Name 2-({6-[bis(carboxymethyl)amino]hexyl}(carboxymethyl)amino)acetic acid
SMILES OC(=O)CN(CCCCCCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Melting Point 236°C
Color White
Quantity 25 g
Formula Weight 348.35
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,6-Diaminohexane-N,N,N′,N′-tetraacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.