Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Diiodohexane, 97+%, stab. with copper

Catalog No. p-7022540
Click to view available options
Quantity:
25 g
100 g

CAS: 629-09-4 | C6H12I2 | 337.97 g/mol

1,6-Diiodohexane is used as a precursor in the preparation of cationic tissue plasminogen activator (t-PA)-containing polyurethane surfaces. It is also used as a solvent additive and in potential power conversion efficiency of polymer or fullerene bulk heterojunction solar cells. Further, it is employed in the preparation of 3-(iodohexyl)-1-(3-(triethoxysilyl)propylcarbamoyl)-1H-benzo[d]imidazol-3-ium iodide and 2,4-dihydroxybenzophenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Diiodohexane is used as a precursor in the preparation of cationic tissue plasminogen activator (t-PA)-containing polyurethane surfaces. It is also used as a solvent additive and in potential power conversion efficiency of polymer or fullerene bulk heterojunction solar cells. Further, it is employed in the preparation of 3-(iodohexyl)-1-(3-(triethoxysilyl)propylcarbamoyl)-1H-benzo[d]imidazol-3-ium iodide and 2,4-dihydroxybenzophenone.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-09-4
Molecular Formula C6H12I2
Molecular Weight (g/mol) 337.97
MDL Number MFCD00001103
InChI Key QLIMAARBRDAYGQ-UHFFFAOYSA-N
Synonym hexamethylene diiodide, hexane, 1,6-diiodo, 1,6-diiodohexane, stabilized with copper, 1,6-di-iodohexane, 1,6-diiodo hexane, pubchem3846, hexamethyleneiodide, 1,6-bis iodanyl hexane, acmc-209tt7, ksc354g6d
PubChem CID 12373
IUPAC Name 1,6-diiodohexane
SMILES ICCCCCCI

Specifications

Density 2.052
Boiling Point 123°C to 128°C (4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5852
Quantity 25 g
Beilstein 1280128
Sensitivity Light sensitive
Solubility Information Immiscible with water.
Formula Weight 337.97
Percent Purity 98%
Chemical Name or Material 1,6-Diiodohexane, stabilized with copper
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-073-5
RTECSNumber MO1700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.