Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Diisocyanatohexane, 99+%

Catalog No. AC115190010
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle

CAS: 822-06-0 | C8H12N2O2 | 168.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 822-06-0
Molecular Formula C8H12N2O2
Molecular Weight (g/mol) 168.20
MDL Number MFCD00002047
InChI Key RRAMGCGOFNQTLD-UHFFFAOYSA-N
Synonym hexamethylene diisocyanate, 1,6-hexamethylene diisocyanate, hmdi, hexane, 1,6-diisocyanato, hexane 1,6-diisocyanate, 1,6-hexylene diisocyanate, hexamethylene-1,6-diisocyanate, 1,6-hexanediol diisocyanate, hdi, isocyanic acid, hexamethylene ester
PubChem CID 13192
ChEBI CHEBI:53578
IUPAC Name 1,6-diisocyanatohexane
SMILES O=C=NCCCCCCN=C=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -67°C
Color Colorless to Yellow
Density 1.0400g/mL
Boiling Point 255.0°C
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4510 to 1.4530
Linear Formula OCN(CH2)6NCO
Quantity 1 kg
Beilstein 04, II, 711
Specific Gravity 1.04
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in benzene,acetone,toluene,,ethyl acetate,chlorobenzene,very soluble in ether
Formula Weight 168.19
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 1, 6-Diisocyanatohexane, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hai
EINECSNumber 212-485-8
RTECSNumber MO1740000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.