Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Diphenyl-1,3,5-hexatriene, Scint, MP Biomedicals

Catalog No. ICN213308
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 1720-32-7
Molecular Formula C18H16
Molecular Weight (g/mol) 232.326
InChI Key BOBLSBAZCVBABY-WPWUJOAOSA-N
Synonym 1,6-diphenyl-1,3,5-hexatriene, 1,6-diphenylhexatriene, diphenylhexatriene, dicinnamyl, 1,6-diphenylhexa-1,3,5-triene, benzene, 1,1'-1,3,5-hexatriene-1,6-diyl bis, 1,3,5-hexatriene, 1,6-diphenyl, 1e,3e,5e-6-phenylhexa-1,3,5-trienyl benzene, 1,1'-1e,3e,5e-hexa-1,3,5-triene-1,6-diyldibenzene
PubChem CID 5376733
ChEBI CHEBI:51594
IUPAC Name [(1E,3E,5E)-6-phenylhexa-1,3,5-trienyl]benzene
SMILES C1=CC=C(C=C1)C=CC=CC=CC2=CC=CC=C2

Specifications

Quantity 1 g
Formula Weight 232.33
Chemical Name or Material 1,6-Diphenyl-1,3,5-Hexatriene, Scint
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.