Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl formate, 98+%, pure

Catalog No. AC150670010
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal drum
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 109-94-4 | C3H6O2 | 74.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-94-4
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.08
MDL Number MFCD00003294
InChI Key WBJINCZRORDGAQ-UHFFFAOYSA-N
Synonym areginal, formic acid, ethyl ester, ethyl methanoate, formic acid ethyl ester, aethylformiat, ethyl formic ester, ethylformate, ethylformiaat, formic ether, ethylformiat
PubChem CID 8025
ChEBI CHEBI:52342
IUPAC Name ethyl formate
SMILES CCOC=O

Specifications

Melting Point -80.0°C
Density 0.9170g/mL
Boiling Point 54.0°C
Flash Point −20°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.3580 to 1.3610
Linear Formula HCO2C2H5
Quantity 1 L
Beilstein 02, 19
Fieser 01,380; 02,197; 04,233; 12,226
Specific Gravity 0.917
Merck Index 15, 3862
Solubility Information Solubility in water: 110g/L (18°C). Other solubilities: freely soluble in ethylacetaat, isobutylacetaat,, butylacetaat, aceton, methylisobutylketon and, methylethylketon, miscible with benzene, ether and alcohol
Viscosity 0.4 mPa.s (20°C)
Formula Weight 74.08
Percent Purity 98+%
Grade Pure
Physical Form Liquid
Chemical Name or Material Ethyl formate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Call a POISON CENTER or doctor/physician if you
EINECSNumber 203-721-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.