Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Hexanediol, 97%

Catalog No. AC120650025
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC120650025 2.5 kg Glass bottle
AC120650010 1 kg Glass bottle
AC120650100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC120650025 Supplier Thermo Scientific Chemicals Supplier No. 120650025
Add to Cart

CAS: 629-11-8 | C6H14O2 | 118.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-11-8
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
InChI Key XXMIOPMDWAUFGU-UHFFFAOYSA-N
Synonym 1,6-hexanediol, hexamethylene glycol, 1,6-dihydroxyhexane, hexamethylenediol, alpha,omega-hexanediol, .alpha.,.omega.-hexanediol, unii-zia319275i, ccris 8982, 6-hydroxy-1-hexanol, 1,6-hexylene glycol
PubChem CID 12374
ChEBI CHEBI:43078
IUPAC Name hexane-1,6-diol
SMILES C(CCCO)CCO

Specifications

Acidity 0.1 mg KOH/g max.
Melting Point 40.0°C to 43.0°C
Color White
Density 0.9600g/mL
Boiling Point 253.0°C to 260.0°C
Flash Point 147°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.457
Linear Formula HO(CH2)6OH
Quantity 2.5 kg
Beilstein 01,484
Merck Index 15,4728
Specific Gravity 0.96
Solubility Information Solubility in water: 500g/L in water. Other solubilities: soluble in alcohol, sparingly soluble in hot ether
Viscosity 30 mPa.s (60°C)
Formula Weight 118.18
Percent Purity 97%
Physical Form Waxy Flakes
Chemical Name or Material 1, 6-Hexanediol
Show More Show Less
EINECSNumber 211-074-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.