Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Hexanedithiol 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
1191-43-1
C6H14S2
150.3
MFCD00004910
SRZXCOWFGPICGA-UHFFFAOYSA-N
1,6-hexanedithiol,1,6-dimercaptohexane,1,6-hexanedimercaptan,usaf uctl-72,hexamethylene dimercaptan,1,6-hexane dithiol,1,6-hexamethylenedithiol,unii-rwn7rm884e,fema no. 3495,hexanedithiol-1,6
14491
hexane-1,6-dithiol
C(CCCS)CCS
1191-43-1
C6H14S2
150.3
MFCD00004910
SRZXCOWFGPICGA-UHFFFAOYSA-N
1,6-hexanedithiol,1,6-dimercaptohexane,1,6-hexanedimercaptan,usaf uctl-72,hexamethylene dimercaptan,1,6-hexane dithiol,1,6-hexamethylenedithiol,unii-rwn7rm884e,fema no. 3495,hexanedithiol-1,6
14491
hexane-1,6-dithiol
C(CCCS)CCS
Specific Gravity 0.99
Boiling Point 237.0°C
Refractive Index 1.5110 to 1.5150
Density 0.9900g/mL
Packaging Glass bottle
Color Colorless to Yellow
Flash Point 90°C
Quantity 5g
Infrared Spectrum Authentic
Melting Point -21.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula HS(CH2)6SH
Beilstein 01, 484
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in fat
Formula Weight 150.3
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 1, 6-Hexanedithiol, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 214-735-1

RTECSNumber : MO3500000

TSCA : TSCA