Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,7-Dihydroxynaphthalene, 97%

Catalog No. p-7030376
Click to view available options
Quantity:
5 g
25 g

CAS: 575-38-2 | C10H8O2 | 160.172 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 575-38-2
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.172
MDL Number MFCD00035720
InChI Key ZUVBIBLYOCVYJU-UHFFFAOYSA-N
Synonym 1,7-dihydroxynaphthalene, 1,7-naphthalenediol, napthalene-1,7-diol, unii-7d42f605cs, ccris 7895, 1,7 dihydroxynaphthalene, 2,8-dihydroxynaphthalene, acmc-1aqz2, unii-m7uz7bfo3a component
PubChem CID 68462
IUPAC Name naphthalene-1,7-diol
SMILES C1=CC2=C(C=C(C=C2)O)C(=C1)O

Specifications

Melting Point 180°C to 184°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 160.17
Percent Purity 97%
Chemical Name or Material 1,7-Dihydroxynaphthalene
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 209-383-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.