Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Cineole, 99%

Catalog No. p-7082034
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA12269AE 100 mL
AAA12269AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA12269AE Supplier Thermo Scientific Chemicals Supplier No. A12269AE
Only null left
Add to Cart

CAS: 470-82-6 | C10H18O | 154.25 g/mol

Eucalyptol is an ingredient in many brands of mouthwash and cough suppressant. It controls airway mucus hypersecretion and asthma via anti-inflammatory cytokine inhibition. Eucalyptol is an effective treatment for nonpurulent rhinosinusitis. Eucalyptol reduces inflammation and pain when applied topically. It kills leukaemia cells in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Eucalyptol is an ingredient in many brands of mouthwash and cough suppressant. It controls airway mucus hypersecretion and asthma via anti-inflammatory cytokine inhibition. Eucalyptol is an effective treatment for nonpurulent rhinosinusitis. Eucalyptol reduces inflammation and pain when applied topically. It kills leukaemia cells in vitro.

Solubility
Soluble in water(3500 mg/L (at 21°C). Miscible with ether, alcohol, chloroform, glacial acetic acid, oils. Soluble in ethanol, ethyl ether; slightly soluble in carbon tetrachloride.

Notes
Store this chemical under refrigerated temperatures and away from mineral acids and bases. Store in a tightly closed container. Store in a dry, well-ventilated area.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 470-82-6
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00167977
InChI Key WEEGYLXZBRQIMU-UHFFFAOYSA-N
Synonym eucalyptol, cineole, 1,8-cineole, 1,8-cineol, cajeputol, 1,8-epoxy-p-menthane, eucalyptole, zineol, eucapur, terpan
PubChem CID 2758
IUPAC Name 2,2,4-trimethyl-3-oxabicyclo[2.2.2]octane
SMILES CC12CCC(CC1)C(C)(C)O2

Specifications

Melting Point 1°C
Density 0.92
Boiling Point 176°C to 177°C
Flash Point 49°C (120°F)
Assay Percent Range 99%
Refractive Index 1.4575
Quantity 100 mL
UN Number UN3271
Beilstein 105109
Merck Index 14,3895
Solubility Information Soluble in water(3500mg/L (at 21°C). Miscible with ether,alcohol,chloroform,glacial acetic acid,oils. Soluble in ethanol,ethyl ether; slightly soluble in carbon tetrachloride.
Formula Weight 154.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Cineole
Show More Show Less
Hazard Category H226-H315-H319
Hazard Statement GHS H Statement
H226
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 207-431-5
RTECSNumber OS9275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.