Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Diaminonaphthalene, 97%

Catalog No. p-2689485
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle

CAS: 479-27-6 | C10H10N2 | 158.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 479-27-6
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.2
MDL Number MFCD00004033
InChI Key YFOOEYJGMMJJLS-UHFFFAOYSA-N
Synonym 1,8-diaminonaphthalene, 1,8-naphthalenediamine, 1,8-naphthylenediamine, unii-ika7029yh9, ccris 8398, 1,8-diaminonapthalene, 1,8-diamino naphthalene, 1,8-naphthalene diamine, acmc-209kb1, ksc236e9t
PubChem CID 68067
IUPAC Name naphthalene-1,8-diamine
SMILES C1=CC2=C(C(=C1)N)C(=CC=C2)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 63.0°C to 66.0°C
Color Brown or Gray to Gray-Red
Boiling Point 205.0°C (12.0 mmHg)
Flash Point 195°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C10H6(NH2)2
Quantity 5 g
Beilstein 13, 205
Merck Index 15, 6457
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether, slightly soluble in chloroform
Formula Weight 158.2
Percent Purity 97%
Physical Form Crystalline Mass or Chunks
Chemical Name or Material 1, 8-Diaminonaphthalene, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If s
EINECSNumber 207-529-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.