Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+%

Catalog No. AC160610025
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle

CAS: 6674-22-2 | C9H16N2 | 152.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6674-22-2
MDL Number MFCD00006930
InChI Key GQHTUMJGOHRCHB-UHFFFAOYSA-N
Synonym 1,8-diazabicyclo 5.4.0 undec-7-ene, 2,3,4,6,7,8,9,10-octahydropyrimido 1,2-a azepine, polycat dbu, dbu, unii-h1ilj6ibux, pyrimido 1,2-a azepine, 2,3,4,6,7,8,9,10-octahydro, 1,8-diazabicyclo 5.4.0-7-undecene, h1ilj6ibux, 1,8-diazabicyclo 5,4,0 undec-7-ene
PubChem CID 81184
IUPAC Name 2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine
SMILES C1CCC2=NCCCN2CC1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -70.0°C
Color Colorless to Yellow
Density 1.0190g/mL
Boiling Point 261.0°C
Flash Point 116°C
Infrared Spectrum Authentic
Assay Percent Range ≥98%
Packaging Glass bottle
Molecular Formula C9H16N2
Refractive Index 1.5210 to 1.5230
Quantity 2.5 kg
Beilstein 23, V,5, 271
Fieser 02,101; 04,119; 05,177; 06,158; 07,87; 11,155; 13,91; 14,109; 15,109; 16,105; 17,99
Merck Index 15, 2996
Solubility Information Solubility in water: soluble.
Molecular Weight (g/mol) 152.241
Formula Weight 152.24
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 1,8-Diazabicyclo[5.4.0]undec-7-ene, ≥98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 229-713-7
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.