Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Diiodooctane, 97+%, stab. with copper

Catalog No. p-7081058
Click to view available options
Quantity:
25 g
50 g

CAS: 24772-63-2 | C8H16I2 | 366.025 g/mol

1,8-Diiodooctane is utilized to improve the power conversion efficiency of polymer solar cells. It is also employed as a processing additive to improve the morphology and the efficiency of bulk heterojunctions solar cells based on the regioregular poly(3-hexylthiophene).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Diiodooctane is utilized to improve the power conversion efficiency of polymer solar cells. It is also employed as a processing additive to improve the morphology and the efficiency of bulk heterojunctions solar cells based on the regioregular poly(3-hexylthiophene).

Solubility
Miscible with methanol.

Notes
Light sensitive. Incompatible with water and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24772-63-2
Molecular Formula C8H16I2
MDL Number MFCD00001106
InChI Key KZDTZHQLABJVLE-UHFFFAOYSA-N
PubChem CID 90605
IUPAC Name 1,8-diiodooctane

Specifications

Density 1.84
Boiling Point 167°C to 169°C (6 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5635
Quantity 25 g
Beilstein 1735437
Sensitivity Light sensitive
Synonym octane, 1,8-diiodo, octamethylene diiodide, pubchem3851, octane,1,8-diiodo, acmc-1cljq, 1,8-bis iodanyl octane, 1,8-diiodooctane, stabilized with copper, stabilizedwithcopperchip, 1,8-diiodooctane, contains copper as stabilizer
Solubility Information Miscible with methanol.
SMILES C(CCCCI)CCCI
Molecular Weight (g/mol) 366.025
Formula Weight 366.03
Percent Purity ≥97%
Chemical Name or Material 1,8-Diiodooctane, Stabilized with copper
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 246-456-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.