Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,8-Naphthalimide, 98%

Click to view available options
Quantity:
10g
50g
250g
81-83-4
C12H7NO2
197.193
MFCD00006920
XJHABGPPCLHLLV-UHFFFAOYSA-N
1,8-naphthalimide,naphthalimide,1h-benz de isoquinoline-1,3 2h-dione,1h-benzo de isoquinoline-1,3 2h-dione,1,8-naphthalenedicarboximide,benzo de isoquinoline-1,3-dione,unii-gi0tv19gbn,naphthalene-1,8-dicarboximide,gi0tv19gbn
66491
benzo[de]isoquinoline-1,3-dione
C1=CC2=C3C(=C1)C(=O)NC(=O)C3=CC=C2
81-83-4
C12H7NO2
197.193
MFCD00006920
XJHABGPPCLHLLV-UHFFFAOYSA-N
1,8-naphthalimide,naphthalimide,1h-benz de isoquinoline-1,3 2h-dione,1h-benzo de isoquinoline-1,3 2h-dione,1,8-naphthalenedicarboximide,benzo de isoquinoline-1,3-dione,unii-gi0tv19gbn,naphthalene-1,8-dicarboximide,gi0tv19gbn
66491
benzo[de]isoquinoline-1,3-dione
C1=CC2=C3C(=C1)C(=O)NC(=O)C3=CC=C2
Applications
It is a useful material for utilization in phosphorescence dye doped organic light emitting diodes (oleds), hole blocking and electron-conducting material.

Solubility
Insoluble in water.

Notes
Store away from and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Melting Point 299°C to 303°C
Quantity 50g
Beilstein 153150
Solubility Information Insoluble in water.
Formula Weight 197.2
Percent Purity 98%
Chemical Name or Material 1,8-Naphthalimide

P264b-P280i-P305+P351+P338

H319

EINECSNumber : 201-379-7

TSCA : Yes

Recommended Storage : Ambient temperatures