Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Nonadiyne, 97%

Catalog No. p-7027207
Click to view available options
Quantity:
5 g
25 g

CAS: 2396-65-8 | C9H12 | 120.195 g/mol

It is used as starting reagent in the synthesis of 2,6-hexadecadiynoic acid, 2,6-nonadecadiynoic acid and 2,9-hexadecadiynoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as starting reagent in the synthesis of 2,6-hexadecadiynoic acid, 2,6-nonadecadiynoic acid and 2,9-hexadecadiynoic acid.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from heavy metal powders and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect from electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2396-65-8
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00008581
InChI Key DMOVPHYFYSASTC-UHFFFAOYSA-N
Synonym 1,8-nonadiyne, acmc-1cak0, 1,8-nonadiyne,, 1,8-nonadiyne 6ci, 7ci, 8ci, 9ci
PubChem CID 16969
IUPAC Name nona-1,8-diyne
SMILES C#CCCCCCC#C

Specifications

Melting Point -21°C
Density 0.799
Boiling Point 55°C to 56°C (13 mmHg)
Flash Point 41°C (105°F)
Odor Characteristic
Refractive Index 1.449
Linear Formula HC≡C(CH2)5C≡CH
Quantity 5 g
UN Number UN3295
Beilstein 1733443
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 120.2
Percent Purity 97%
Chemical Name or Material 1,8-Nonadiyne
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 219-254-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.