Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Octanediol, 98+%

Catalog No. p-7022566
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 629-41-4 | C8H18O2 | 146.23 g/mol

1,8-Octanediol is used in cosmetics, perfumes, ink, essence and in UV coating. It acts as a precursor in the synthesis of polymer and in the manufacture of pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Octanediol is used in cosmetics, perfumes, ink, essence and in UV coating. It acts as a precursor in the synthesis of polymer and in the manufacture of pharmaceuticals.

Solubility
Soluble in water and methanol.

Notes
Incompatible with strong oxidizing agents, reducing agents, acid chlorides acid, anhydrides and chloroformates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-41-4
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00002989
InChI Key OEIJHBUUFURJLI-UHFFFAOYSA-N
PubChem CID 69420
ChEBI CHEBI:44630
IUPAC Name octane-1,8-diol
SMILES C(CCCCO)CCCO

Specifications

Melting Point 58°C to 61°C
Boiling Point 171°C to 173°C (20 mm)
Flash Point 148°C (298°F)
Assay Percent Range ≥98%
Linear Formula HO(CH2)8OH
Quantity 25 g
Beilstein 1633499
Synonym 1,8-octanediol, octamethylene glycol, 1,8-dihydroxyoctane, octan-1,8-diol, 1,8-octandiol, 1, 8-octandiol, octane-1,8-diol;, acmc-209nan, ksc358i9p
Solubility Information Soluble in water and methanol.
Formula Weight 146.23
Percent Purity ≥98%
Chemical Name or Material 1, 8-Octanediol
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-090-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.