Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,9-Decadiene, 97%

Catalog No. AA3120718
Click to view available options
Quantity:
10 g
50 g

CAS: 1647-16-1 | C10H18 | 138.254 g/mol

It is widely used in the manufacturers and suppliers of pharmaceuticals and intermediates in the preparation of fine chemicals. 1,9-decadiene is also used as a starting material in ADMET polymerizations .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1647-16-1
Molecular Formula C10H18
Molecular Weight (g/mol) 138.254
MDL Number MFCD00008670
InChI Key NLDGJRWPPOSWLC-UHFFFAOYSA-N
Synonym 1,9-decadiene, decadiene-1,9, unii-2kwz01g244, acmc-1c2oy, dsstox_cid_2159, dsstox_rid_76506, dsstox_gsid_22159, chembl31020
PubChem CID 15439
IUPAC Name deca-1,9-diene
SMILES C=CCCCCCCC=C

Specifications

Density 0.75 g/mL
Boiling Point 169°C
Flash Point 42°C (107°F)
Refractive Index 1.4325
Quantity 50 g
UN Number UN3295
Solubility Information It is soluble in organic solvents.
Formula Weight 138.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1,9-Decadiene
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 216-711-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.