Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,9-Nonanedicarboxylic Acid 97.0+%, TCI America™
SDP

Catalog No. N033325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N033325G 25 g
N03335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N033325G Supplier TCI America Supplier No. N033325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1852-04-6
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.28
MDL Number MFCD00004444
InChI Key LWBHHRRTOZQPDM-UHFFFAOYSA-N
Synonym 1,9-nonanedicarboxylic acid, 1,11-undecanedioic acid, undecanedionic acid, 1,9-nonanedicarboxylicacid, undecandioic acid, hendecanedioic acid, unii-7059gfk8nv, nonamethylenedicarboxylic acid, hendecanedioate, undecanedioate
PubChem CID 15816
ChEBI CHEBI:73713
IUPAC Name undecanedioic acid
SMILES OC(=O)CCCCCCCCCC(O)=O

Specifications

Melting Point 111°C
Color White-Yellow
Quantity 25 g
Formula Weight 216.28
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,9-Nonanedicarboxylic Acid
EINECSNumber (2)-0878
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.