Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,9-Nonanediol, 98%

Catalog No. p-175484
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

Catalog No. AC129120250

Supplier: Thermo Scientific Chemicals 129120250

Only null left
Add to Cart

CAS: 3937-56-2 | C9H20O2 | 160.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3937-56-2
Molecular Formula C9H20O2
Molecular Weight (g/mol) 160.26
MDL Number MFCD00002991
InChI Key ALVZNPYWJMLXKV-UHFFFAOYSA-N
Synonym 1,9-nonanediol, 1,9-dihydroxynonane, .alpha.,.omega.-nonanediol, nonamethylene glycol, alpha,omega-nonanediol, 1,9-nonandiol, 1,9-nonane-diol, nonan-1,9-diol, acmc-209j4x
PubChem CID 19835
IUPAC Name nonane-1,9-diol
SMILES C(CCCCO)CCCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 44°C to 48°C
Color White
Density 0.9180g/mL
Boiling Point 177°C (15.0 mmHg)
Flash Point 162°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula HO(CH2)9OH
Quantity 25 g
Beilstein 01, 493
Specific Gravity 0.918
Solubility Information Solubility in water: 9g/L (20°C)
Viscosity 52.6 mPa.s (50°C)
Formula Weight 160.26
Percent Purity 99%
Physical Form Fused Crystalline Solid
Chemical Name or Material 1, 9-Nonanediol, 99%
Show More Show Less
EINECSNumber 223-517-5
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.