Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Acetylimidazole, 98%, Thermo Scientific Chemicals

Catalog No. AC169500050
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2466-76-4
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD00005287
InChI Key VIHYIVKEECZGOU-UHFFFAOYSA-N
Synonym 1-acetylimidazole, n-acetylimidazole, 1-1h-imidazol-1-yl ethanone, 1h-imidazole, 1-acetyl, acetylimidazole, 1-acetyl-1h-imidazole, imidazole, 1-acetyl, 3-acetylimidazole, unii-zmp8x1y11g, ccris 6534
PubChem CID 17174
ChEBI CHEBI:16984
IUPAC Name 1-imidazol-1-ylethanone
SMILES CC(=O)N1C=CN=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 93°C to 96°C
Color Beige to White or Tan
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,ether,chloroform,thf
Formula Weight 110.12
Percent Purity 98%
Physical Form Adhering Crystals or Crystalline Powder
Chemical Name or Material 1-Acetylimidazole, 98%
Show More Show Less
Hazard Category MOISTURE SENSITIVE
EINECSNumber 219-577-7
RTECSNumber NI3400000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.