Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Adamantanamine Hydrochloride 99.0+%, TCI America™
SDP

Catalog No. a058825g Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
250 g

Chemical Identifiers

CAS 665-66-7
Molecular Formula C10H18ClN
Molecular Weight (g/mol) 187.71
MDL Number MFCD00074723
InChI Key WOLHOYHSEKDWQH-UHFFFAOYSA-N
Synonym amantadine hydrochloride, 1-adamantanamine hydrochloride, symmetrel, 1-aminoadamantane hydrochloride, amantadine hcl, adamantanamine hydrochloride, amazolon, midantan, midantane, mydantane
PubChem CID 64150
ChEBI CHEBI:2619
IUPAC Name hydrogen adamantan-1-amine chloride
SMILES [H+].[Cl-].NC12CC3CC(CC(C3)C1)C2

Specifications

Melting Point 360°C
Color White
Quantity 25 g
Formula Weight 187.71
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Adamantanamine Hydrochloride
RTECSNumber AU4375000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.